Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butylcyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. B193025G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 70568-60-4
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD01321147
InChI Key NUANGSLQWFBVEH-UHFFFAOYSA-N
PubChem CID 4132186
IUPAC Name 4-butylcyclohexan-1-ol
SMILES CCCCC1CCC(CC1)O

Specifications

Color Colorless
Boiling Point 122°C
Quantity 25 g
Formula Weight 156.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Butylcyclohexanol (cis- and trans- mixture)
EINECSNumber (3)-2301
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.