Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Carboxy-2-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193740
Click to view available options
Quantity:
1 g
5 g

Specifications

Chemical Name or Material 4-Carboxy-2-fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 851335-07-4
Color White
Molecular Formula C7H6BFO4
Quantity 1 g
MDL Number MFCD08458473
Synonym 4-Carboxy-2-fluorobenzeneboronic Acid
InChI Key TVNVDRMGOOLVOX-UHFFFAOYSA-N
SMILES OB(O)C1=C(F)C=C(C=C1)C(O)=O
IUPAC Name 4-(dihydroxyboranyl)-3-fluorobenzoic acid
Molecular Weight (g/mol) 183.93
PubChem CID 23134177
Formula Weight 183.93
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.