Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Carboxy-3-chlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C25235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2523-5G 5 g
C25231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2523-5G Supplier TCI America Supplier No. C25235G
Add to Cart
Add to Cart

Specifications

CAS 136496-72-5
Melting Point 232°C
Color White
Molecular Formula C7H6BClO4
Quantity 5 g
MDL Number MFCD06656268
Synonym 4-carboxy-3-chlorophenylboronic acid, 3-chloro-4-carboxyphenylboronic acid, 4-carboxy-3-chlorobenzeneboronic acid, 2-chloro-4-dihydroxyboranyl benzoic acid, 4-carboxy-3-chlorobenzeneboronicacid
InChI Key QFAFGWXQNDYXPZ-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)C(=O)O)Cl)(O)O
IUPAC Name 4-borono-2-chlorobenzoic acid
Molecular Weight (g/mol) 200.381
PubChem CID 14903223
Formula Weight 200.38
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.