Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tert-Butyl 3-(2-aminoethyl)azetidine-1-carboxylate, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
| Assay Percent Range | 94% min. (GC) |
| Quantity | 1 g |
| Formula Weight | 200.28 |
| Percent Purity | 95% |
| Chemical Name or Material | tert-Butyl 3-(2-aminoethyl)azetidine-1-carboxylate |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction