Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Carboxyphenylboronic acid, 97%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
Specifications
Melting Point | 238.0°C to 240.0°C |
Infrared Spectrum | Authentic |
Quantity | 25g |
Packaging | Glass bottle |
Color | White to Beige |
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Linear Formula | HOOCC6H4B(OH)2 |
Solubility Information | Solubility in water: 25g/L. |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning