Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-1-methylpiperidine hydrochloride, 97%, Thermo Scientific™

Catalog No. AC109491000
Click to view available options
:
100g; Glass bottle

Specifications

CAS 5382-23-0
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H13Cl2N
Quantity 100g
MDL Number MFCD00012834
Synonym 4-chloro-1-methylpiperidine hydrochloride, 1-methyl-4-chloropiperidine hydrochloride, 4-chloro-n-methylpiperidine hydrochloride, 4-chloro-1-methylpiperidinium chloride, piperidine, 4-chloro-1-methyl-, hydrochloride, n-methyl-4-chloropiperidine hydrochloride, 1-methyl-4-chloro piperidine, pubchem2461, acmc-1apdo, ksc495i8n
InChI Key QDOJNWFOCOWAPO-UHFFFAOYSA-N
SMILES CN1CCC(CC1)Cl.Cl
IUPAC Name 4-chloro-1-methylpiperidine;hydrochloride
Molecular Weight (g/mol) 170.077
PubChem CID 3034158
Formula Weight 170.08
Percent Purity ≥96.0%
Chemical Name or Material 4-Chloro-1-methylpiperidine hydrochloride, 97%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 226-375-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.