Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2,5-dimethoxynitrobenzene 98.0+%, TCI America™
SDP

Catalog No. C1244250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1244250G 250 g
C124425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1244250G Supplier TCI America Supplier No. C1244250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6940-53-0
Molecular Formula C8H8ClNO4
Molecular Weight (g/mol) 217.605
MDL Number MFCD00024577
InChI Key ORLPGMKKCAEWOW-UHFFFAOYSA-N
PubChem CID 81362
IUPAC Name 1-chloro-2,5-dimethoxy-4-nitrobenzene
SMILES COC1=CC(=C(C=C1[N+](=O)[O-])OC)Cl

Specifications

Melting Point 141°C
Quantity 250 g
Formula Weight 217.61
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2,5-dimethoxynitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.