Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2,6-dimethyl-3-nitropyridine, 98%, Thermo Scientific™

Catalog No. AAH6418003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6418003 1 g
AAH64180MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6418003 Supplier Thermo Scientific Chemicals Supplier No. H6418003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15513-48-1
Molecular Formula C7H7ClN2O2
Molecular Weight (g/mol) 186.595
MDL Number MFCD08543468
InChI Key PSBGFLWVQDGETK-UHFFFAOYSA-N
Synonym pyridine, 4-chloro-2,6-dimethyl-3-nitro, 4-chloro-3-nitro-2,6-lutidine, pubchem11177, 4-chloranyl-2,6-dimethyl-3-nitro-pyridine
PubChem CID 10702725
IUPAC Name 4-chloro-2,6-dimethyl-3-nitropyridine
SMILES CC1=NC(=C(C(=C1)Cl)[N+](=O)[O-])C

Specifications

Melting Point 70°C to 73°C
Color White
Quantity 1 g
Beilstein 1569403
Formula Weight 186.6
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Chloro-2,6-dimethyl-3-nitropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.