Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-fluoro-1-iodobenzene 98.0+%, TCI America™
SDP

Catalog No. C149925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C149925G 25 g
C14995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C149925G Supplier TCI America Supplier No. C149925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6797-79-1
Molecular Formula C6H3ClFI
Molecular Weight (g/mol) 256.44
MDL Number MFCD00079729
InChI Key RSTFBOIFYXJIMR-UHFFFAOYSA-N
Synonym 4-chloro-2-fluoroiodobenzene, 2-fluoro-4-chloroiodobenzene, 1-chloro-3-fluoro-4-iodobenzene, 4-chloro-2-fluoro-1-iodo-benzene, 4-chloro-2-fluoro-iodobenzene, pubchem3435, acmc-1b71k, 2-iodo-5-chlorofluorobenzene, 2-iodo-5-chlorofluoro benzene, 3-fluoro-4-iodophenyl chloride
PubChem CID 2736562
IUPAC Name 4-chloro-2-fluoro-1-iodobenzene
SMILES FC1=C(I)C=CC(Cl)=C1

Specifications

Quantity 25 g
Formula Weight 256.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-fluoro-1-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.