Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Chloro-2'-fluoroacetanilide 98.0+%, TCI America™
SDP

Catalog No. C169225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C169225G 25 g
C16925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C169225G Supplier TCI America Supplier No. C169225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59280-70-5
Molecular Formula C8H7ClFNO
Molecular Weight (g/mol) 187.598
MDL Number MFCD00042520
InChI Key GVRKNSAEOVXHOS-UHFFFAOYSA-N
Synonym 4'-chloro-2'-fluoroacetanilide, n-4-chloro-2-fluorophenyl acetamide, 4-chloro-2-fluoroacetanilide, 2-fluoro-4-chloroacetanilide, 2'-fluoro-4'-chloroacetanilide, acetamide, n-4-chloro-2-fluorophenyl, acmc-1az4m, 4-chloro2-fluoroacetanilide, 4'-chloro-2'-fluoroacetanilid, 4'-chloro-2'-fluroacetanilide
PubChem CID 853174
IUPAC Name N-(4-chloro-2-fluorophenyl)acetamide
SMILES CC(=O)NC1=C(C=C(C=C1)Cl)F

Specifications

Melting Point 157°C
Quantity 25 g
Formula Weight 187.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloro-2′-fluoroacetanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.