Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193786 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Specifications

Chemical Name or Material 4-Chloro-2-fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 160591-91-3
Melting Point 250°C
Color White-Yellow
Molecular Formula C6H5BClFO2
Quantity 1 g
MDL Number MFCD02684293
Synonym 4-chloro-2-fluorobenzeneboronic acid, 4-chloro-2-fluorophenyl boronic acid, 4-chloro-2-fluorophenylboronicacid, 2-fluoro-4-chlorophenylboronic acid, b-4-chloro-2-fluorophenyl-boronic acid, 4-chloro-2-fluorophenyl-boronic acid, boronic acid, 4-chloro-2-fluorophenyl, boronic acid, b-4-chloro-2-fluorophenyl, pubchem5034, 4-chloro-2-fluoro-phenyl boronic acid
InChI Key YBNDRTRLXPEWKQ-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)Cl)F)(O)O
IUPAC Name (4-chloro-2-fluorophenyl)boronic acid
Molecular Weight (g/mol) 174.362
PubChem CID 4193595
Formula Weight 174.36
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.