Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-hydroxypyridine 98.0+%, TCI America™
SDP

Catalog No. c31201g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 40673-25-4
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.54
MDL Number MFCD03093022
InChI Key OZUXGFRLSKQVMI-UHFFFAOYSA-N
Synonym 4-chloro-2-hydroxypyridine, 4-chloropyridin-2-ol, 4-chloro-2-pyridinol, 2 1h-pyridinone, 4-chloro, 4-chloro-2 1h-pyridinone, 4-chloro-2 1h-pyridone, 4-chloropyridone, 4-chloro pyridone, 4-chloro-2-pyridone, pubchem9621
PubChem CID 2757611
IUPAC Name 4-chloro-1,2-dihydropyridin-2-one
SMILES ClC1=CC(=O)NC=C1

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 1 g
Formula Weight 129.54
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-hydroxypyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.