Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-iodoaniline, 98+%, Thermo Scientific Chemicals

Catalog No. AC396840050
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63069-48-7
MDL Number MFCD01863737
InChI Key FLEJOBRWKBPUOX-UHFFFAOYSA-N
Synonym 2-iodo-4-chloroaniline, 4-chloro-2-iodo-aniline, 4-chloro-2-iodo-phenylamine, 4-chloro-2-iodophenylamine, benzenamine, 4-chloro-2-iodo, zlchem 532, pubchem3645, 4-chloro-6-iodoaniline, acmc-1bcyk, 2-iodo-4-chloro-aniline
PubChem CID 2757615
IUPAC Name 4-chloro-2-iodoaniline
SMILES C1=CC(=C(C=C1Cl)I)N

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 39.0°C to 43.0°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Molecular Formula C6H5ClIN
Linear Formula ClC6H3(I)NH2
Quantity 5 g
Molecular Weight (g/mol) 253.467
Formula Weight 253.47
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 4-chloro-2-iodoaniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.