Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193870
Click to view available options
Quantity:
1 g
5 g

Specifications

Chemical Name or Material 4-Chloro-2-methylphenylboronic Acid (contains varying amounts of Anhydride)
Melting Point 195°C
CAS 209919-30-2
Color White-Yellow
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C7H8BClO2
MDL Number MFCD02683107
Synonym 4-chloro-2-methylbenzeneboronic acid, 4-chloro-2-methylphenyl boronic acid, 4-chloro-o-tolylboronic acid, 4-chloro-o-tolueneboronic acid, 4-chloro-2-methyl-phenyl boronic acid, 2-methyl-4-chlorophenylboronic acid, 2-methyl-4-chloro-phenylboronic acid, 2-methyl-4-chlorophenyl-boronic acid, boronic acid, 4-chloro-2-methylphenyl
InChI Key SRXXSLUUAWHGBZ-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)Cl)C)(O)O
IUPAC Name (4-chloro-2-methylphenyl)boronic acid
Molecular Weight (g/mol) 170.399
PubChem CID 5017600
Formula Weight 170.40
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.