Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-methylpyrimidine 98.0+%, TCI America™
SDP

Catalog No. C28351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28351G 1 g
C2835200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28351G Supplier TCI America Supplier No. C28351G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4994-86-9
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD00234069
InChI Key WDTVJRYCMIZPMX-UHFFFAOYSA-N
PubChem CID 424447
IUPAC Name 4-chloro-2-methylpyrimidine
SMILES CC1=NC=CC(=N1)Cl

Specifications

Melting Point 57°C
Color White-Yellow
Boiling Point 168°C
Quantity 1 g
Formula Weight 128.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-methylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.