Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Chloro-2-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Chemical Name or Material | 4-Chloro-2-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride) |
CAS | 313545-41-4 |
Melting Point | 182°C |
Color | White-Yellow |
Molecular Formula | C7H5BClF3O2 |
Quantity | 1 g |
MDL Number | MFCD04973086 |
Synonym | 4-chloro-2-trifluoromethyl phenylboronic acid, 4-chloro-2-trifluoromethylphenylboronic acid, 4-chloro-2-trifluoromethyl benzeneboronic acid, 4-chloro-2-trifluoromethyl phenyl boronic acid, 2-chloro-4-trifluoromethyl-phenylboronic acid, boronic acid, 4-chloro-2-trifluoromethyl phenyl, 4-chloro-2-trifluoromethyl phenylboronicacid, pubchem5158, acmc-20aic7 |
InChI Key | YAPBOBGBYQQYHX-UHFFFAOYSA-N |
SMILES | B(C1=C(C=C(C=C1)Cl)C(F)(F)F)(O)O |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction