Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3,5-dimethylphenol, 98+%

Catalog No. p-7026136
Click to view available options
Quantity:
100 g
500 g

CAS: 88-04-0 | C8H9ClO | 156.609 g/mol

It is used in hospitals and households for disinfection and sanitation. It is used in wound-cleansing applications and household antiseptics such as Dettol liquid, cream and ointments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in hospitals and households for disinfection and sanitation. It is used in wound-cleansing applications and household antiseptics such as Dettol liquid, cream and ointments.

Solubility
Soluble in benzene, toluene, glycerin, chloroform, acetone, isopropanol, alcohol, ether, terpenes and in solutions of alkali hydroxides. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-04-0
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00002324
InChI Key OSDLLIBGSJNGJE-UHFFFAOYSA-N
Synonym chloroxylenol, dettol, 4-chloro-3,5-xylenol, pcmx, benzytol, p-chloro-m-xylenol, 2-chloro-m-xylenol, 4-chloro-m-xylenol, ottasept, desson
PubChem CID 2723
ChEBI CHEBI:34393
IUPAC Name 4-chloro-3,5-dimethylphenol
SMILES CC1=CC(=CC(=C1Cl)C)O

Specifications

Melting Point 112°C to 116°C
Boiling Point ∼246°C
Flash Point 138°C (280°F)
Odor Odorless
Quantity 100 g
Beilstein 1862539
Merck Index 14,2176
Solubility Information Soluble in benzene,toluene,glycerin,chloroform,acetone,isopropanol,alcohol,ether,terpenes and in solutions of alkali hydroxides. Slightly soluble in water.
Formula Weight 156.61
Percent Purity ≥98%
Chemical Name or Material 4-Chloro-3,5-dimethylphenol
Show More Show Less
Hazard Category H302-H315-H317-H319
Hazard Statement GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 201-793-8
RTECSNumber ZE6850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.