Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-fluoroaniline 97.0+%, TCI America™
SDP

Catalog No. C251225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C251225G 25 g
C25125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C251225G Supplier TCI America Supplier No. C251225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 367-22-6
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD01090987
InChI Key ACMJJQYSPUPMPN-UHFFFAOYSA-N
Synonym 3-fluoro-4-chloroaniline, 4-chloro-3-fluoroanline, 4-chloro-3-fluoro-phenylamine, benzenamine, 4-chloro-3-fluoro, 4-chloro-3-fluorophenylamine, pubchem1516, acmc-1co9a, 4-chloro-3-fluoro-aniline, 4-chloro-3-fluorobenzenamine, ksc226g4r
PubChem CID 2736511
IUPAC Name 4-chloro-3-fluoroaniline
SMILES NC1=CC=C(Cl)C(F)=C1

Specifications

Melting Point 60°C
Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 2811
Boiling Point 226°C
Formula Weight 145.56
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material 4-Chloro-3-fluoroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.