Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193907
Click to view available options
Quantity:
1 g
5 g

Specifications

Chemical Name or Material 4-Chloro-3-fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 137504-86-0
Melting Point 144°C
Color White
Molecular Formula C6H5BClFO2
Quantity 1 g
MDL Number MFCD01319010
Synonym 4-chloro-3-fluorophenyl boronic acid, 4-chloro-3-fluorobenzeneboronic acid, 3-fluoro-4-chlorophenylboronic acid, 4-chloro-3-fluoro-phenyl boronic acid, boronic acid, 4-chloro-3-fluorophenyl, 4-chloro-3-fluorobenzeneboronicacid, pubchem1776, ksc174k0r, acmc-1c195, 4-chloro3-fluorophenylboronic acid
InChI Key CMJQIHGBUKZEHP-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)Cl)F)(O)O
IUPAC Name (4-chloro-3-fluorophenyl)boronic acid
Molecular Weight (g/mol) 174.362
PubChem CID 2778656
Formula Weight 174.36
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.