Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-3-iodophenol, 98%

Click to view available options
Quantity:
1g
10g
202982-72-7
C6H4ClIO
254.451
MFCD00070769
UPIATGHEVZKVRT-UHFFFAOYSA-N
phenol, 4-chloro-3-iodo,acmc-20adn2,4-chloro-3-iodo-phenol,phenol,4-chloro-3-iodo,4-chloro-3-iodophenol
22928729
4-chloro-3-iodophenol
C1=CC(=C(C=C1O)I)Cl
202982-72-7
C6H4ClIO
254.451
MFCD00070769
UPIATGHEVZKVRT-UHFFFAOYSA-N
phenol, 4-chloro-3-iodo,acmc-20adn2,4-chloro-3-iodo-phenol,phenol,4-chloro-3-iodo,4-chloro-3-iodophenol
22928729
4-chloro-3-iodophenol
C1=CC(=C(C=C1O)I)Cl
Melting Point 75°C to 77°C
Quantity 1g
Formula Weight 254.45
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-iodophenol

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger