Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-methylbenzoic acid, 98%

Catalog No. AAL0853309
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0853309 10 g
AAL0853304 2 g
AAL0853318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0853309 Supplier Thermo Scientific Chemicals Supplier No. L0853309
Add to Cart

CAS: 7697-29-2 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-29-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00045853
InChI Key MRUKIIWRMSYKML-UHFFFAOYSA-N
Synonym 4-chloro-m-toluic acid, 4-chloro-3-methyl-benzoic acid, 4-chloro-3-methyl benzoic acid, benzoic acid, 4-chloro-3-methyl, m-toluic acid, 4-chloro, pubchem3666, 5-carboxy-2-chlorotoluene, acmc-1bg77, ksc497q5r, 4-chloranyl-3-methyl-benzoic acid
PubChem CID 282989
IUPAC Name 4-chloro-3-methylbenzoic acid
SMILES CC1=CC(=CC=C1Cl)C(O)=O

Specifications

Melting Point 212°C to 216°C
Quantity 10 g
Beilstein 1936535
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-methylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.