Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-4'-hydroxydiphenyl Ether 95.0+%, TCI America™
SDP

Catalog No. C233525G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Specifications

CAS 21567-18-0
Melting Point 83°C
Boiling Point 169°C
Molecular Formula C12H9ClO2
Quantity 25 g
MDL Number MFCD00125839
Synonym 4-(4-Chlorophenoxy)phenol
InChI Key XQMRZWSYBUCVAX-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1O)OC2=CC=C(C=C2)Cl
IUPAC Name 4-(4-chlorophenoxy)phenol
Molecular Weight (g/mol) 220.652
PubChem CID 12346120
Formula Weight 220.65
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-4′-hydroxydiphenyl Ether
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.