Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-chloro-5-methyl-6-phenylthieno[2,3-d]pyrimidine, 97%, Thermo Scientific™

Catalog No. AW01194DA
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AW01194DA 1 g
AW01194EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AW01194DA Supplier Thermo Scientific Chemicals Supplier No. AW01194DA
Add to Cart

Chemical Identifiers

CAS 306934-78-1
Molecular Formula C13H9ClN2S
Molecular Weight (g/mol) 260.74
MDL Number MFCD02179828
InChI Key LRTKDTZXCFGPEU-UHFFFAOYSA-N
Synonym 4-chloro-5-methyl-6-phenylthieno 2,3-d pyrimidine, 4-chloro-5-methyl-6-phenylthiopheno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-methyl-6-phenyl, 4-chloranyl-5-methyl-6-phenyl-thieno 2,3-d pyrimidine
PubChem CID 2735772
IUPAC Name 4-chloro-5-methyl-6-phenylthieno[2,3-d]pyrimidine
SMILES CC1=C(SC2=NC=NC(Cl)=C12)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 260.75
Percent Purity 97%
Chemical Name or Material 4-chloro-5-methyl-6-phenylthieno[2, 3-d]pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.