Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-6,7-dihydro-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidine, 96%

Catalog No. AAH3220803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3220803 1 g
AAH3220806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3220803 Supplier Thermo Scientific Chemicals Supplier No. H3220803
Add to Cart

CAS: 300816-22-2 | C9H7ClN2S | 210.68 g/mol

Used in the synthesis of some novel thieno [2, 3-d] pyrimidines and their antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of some novel thieno [2, 3-d] pyrimidines and their antibacterial activity.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 300816-22-2
Molecular Formula C9H7ClN2S
Molecular Weight (g/mol) 210.68
MDL Number MFCD00572400
InChI Key JKXJMJPJOYMQSK-UHFFFAOYSA-N
PubChem CID 789817
IUPAC Name 1-chloro-7,8-dihydro-6H-cyclopenta[2,3]thieno[2,4-d]pyrimidine
SMILES ClC1=C2C3=C(CCC3)SC2=NC=N1

Specifications

Melting Point 104°C to 105°C
Quantity 1 g
Synonym 4-chloro-6,7-dihydro-5h-cyclopenta 4,5 thieno 2,3-d pyrimidine, 4-chloro-2,3-dihydro-1h-8-thia-5,7-diaza-cyclopenta a indene, 5h-cyclopenta 4,5 thieno 2,3-d pyrimidine,4-chloro-6,7-dihydro, 12-chloro-7-thia-9,11-diazatricyclo 6.4.0.0?,? dodeca-1 12 ,2 6 ,8,10-tetraene, 4-chloro-5,6,7-trihydrocyclopenta 1,2-d pyrimidino 4,5-b thiophene, 12-chloro-7-thia-9,11-diazatricyclo 6.4.0.0 2 ,? dodeca-1 12 ,2 6 ,8,10-tetraene, 4-chloro-6,7-dihydro-5h-cyclopenta 4.5 thieno 2.3-d pyrimidine, 5h-cyclopenta 4,5 thieno 2,3-d pyrimidine, 4-chloro-6,7-dihydro, 12-chloro-7-thia-9,11-diazatricyclo 6.4.0.0 2 , dodeca-1 12 ,2 6 ,8,10-tetraene
Formula Weight 210.69
Percent Purity 96%
Chemical Name or Material 4-Chloro-6,7-dihydro-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.