Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-N-(3-chloro-2-methylphenyl)butyramide, 97%, Thermo Scientific™

Catalog No. AAH5943903
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500346-91-8
Molecular Formula C11H13Cl2NO
Molecular Weight (g/mol) 246.131
MDL Number MFCD01358154
InChI Key YTFOKDALOHUPSO-UHFFFAOYSA-N
Synonym 4-chloro-n-3-chloro-2-methylphenyl butanamide, 4-chloro-n-3-chloro-2-methylphenyl butyramide
PubChem CID 237738
IUPAC Name 4-chloro-N-(3-chloro-2-methylphenyl)butanamide
SMILES CC1=C(C=CC=C1Cl)NC(=O)CCCCl

Specifications

Quantity 1 g
Formula Weight 246.13
Percent Purity 97%
Chemical Name or Material 4-Chloro-N-(3-chloro-2-methylphenyl)butyramide
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.