Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-N-(4-methoxyphenyl)benzamide, 97%

Catalog No. AAH5546903
Click to view available options
Quantity:
250 mg
1 g

CAS: 4018-82-0 | C14H12ClNO2 | 261.705 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4018-82-0
Molecular Formula C14H12ClNO2
Molecular Weight (g/mol) 261.705
MDL Number MFCD00508611
InChI Key UVGPQLMHIRNFEE-UHFFFAOYSA-N
Synonym 4-chloro-n-4-methoxyphenyl benzamide, n-p-chlorobenzoyl-p-anisidine, 4-chloro-n-4-methoxy-phenyl-benzamide, benzamide, 4-chloro-n-4-methoxyphenyl, cambridge id 5190257, n-4-methoxyphenyl-4-chlorobenzamide, 4-chloro-n-4-methoxyphenyl-benzamide, 4-chloro-n-4-methoxyphenyl benzamide #, 4-chlorobenzenamide, n-4-methoxyphenyl
PubChem CID 221848
IUPAC Name 4-chloro-N-(4-methoxyphenyl)benzamide
SMILES COC1=CC=C(C=C1)NC(=O)C2=CC=C(C=C2)Cl

Specifications

Quantity 1 g
Formula Weight 261.71
Percent Purity 97%
Chemical Name or Material 4-Chloro-N-(4-methoxyphenyl)benzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.