Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-N-cyclohexyl-3-nitrobenzamide, 97%, Thermo Scientific™

Catalog No. AAH59836MD
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 321531-75-3
Molecular Formula C13H15ClN2O3
Molecular Weight (g/mol) 282.724
MDL Number MFCD01182372
InChI Key NXILMCJKLLFJES-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrophenyl-n-cyclohexylcarboxamide
PubChem CID 711288
IUPAC Name 4-chloro-N-cyclohexyl-3-nitrobenzamide
SMILES C1CCC(CC1)NC(=O)C2=CC(=C(C=C2)Cl)[N+](=O)[O-]

Specifications

Quantity 250 mg
Formula Weight 282.72
Percent Purity 97%
Chemical Name or Material 4-Chloro-N-cyclohexyl-3-nitrobenzamide
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.