Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4'-Chloroacetoacetanilide, 98%, Thermo Scientific™
Click to view available options
:
5g; Glass bottle
Specifications
Specifications
CAS | 101-92-8 |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Molecular Formula | C10H10ClNO2 |
Linear Formula | CH3COCH2CONHC6H4Cl |
Quantity | 5g |
MDL Number | MFCD00000613 |
Show More |
Safety and Handling
EINECSNumber | 202-989-6 |
RTECSNumber | AK4375000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction