Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Chloroacetophenone, 98+%

Catalog No. p-85387
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 99-91-2 | C8H7ClO | 154.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-91-2
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000624
InChI Key BUZYGTVTZYSBCU-UHFFFAOYSA-N
Synonym 4'-chloroacetophenone, 1-4-chlorophenyl ethanone, 4-chloroacetophenone, p-chloroacetophenone, ethanone, 1-4-chlorophenyl, acetophenone, 4'-chloro, 1-4-chlorophenyl ethan-1-one, 4-acetylchlorobenzene, p-chloracetophenone, p-acetylchlorobenzene
PubChem CID 7467
ChEBI CHEBI:27538
IUPAC Name 1-(4-chlorophenyl)ethanone
SMILES CC(=O)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 18.0°C
Color Colorless to Yellow
Density 1.1900g/mL
Boiling Point 232.0°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5539 to 1.5559
Linear Formula ClC6H4COCH3
Quantity 5 g
Beilstein 07,281
Merck Index 15,2116
Specific Gravity 1.19
Solubility Information Solubility in water: 111 mg/l (25°C).
Formula Weight 154.6
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material 4'-Chloroacetophenone, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rin
EINECSNumber 202-800-7
RTECSNumber KM5600000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.