Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloroaniline, 98%

Catalog No. p-85450
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
Packaging:
Glass bottle

Catalog No. AC108590050

Supplier: Thermo Scientific Chemicals 108590050

Only null left
Add to Cart

CAS: 106-47-8 | C6H6ClN | 127.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-47-8
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00007835
InChI Key QSNSCYSYFYORTR-UHFFFAOYSA-N
Synonym p-chloroaniline, 4-chlorobenzenamine, p-chloraniline, benzenamine, 4-chloro, p-chlorophenylamine, 4-chlorophenylamine, para-chloroaniline, 1-amino-4-chlorobenzene, p-aminochlorobenzene, 4-aminochlorobenzene
PubChem CID 7812
ChEBI CHEBI:20331
IUPAC Name 4-chloroaniline
SMILES C1=CC(=CC=C1N)Cl

Specifications

Melting Point 68.0°C to 72.0°C
Color Beige to Brown-Purple
CAS Min % 97.5
CAS Max % 100.0
Boiling Point 232.0°C
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Solid
Assay Percent Range 97.5% min. (GC)
Linear Formula ClC6H4NH2
Viscosity 1.2 mPa.s (90°C)
Beilstein 12,607
Merck Index 15,2118
Solubility Information Solubility in water: 3g/L in water (20°C). Other solubilities: freely soluble in alcohol, ether, acetone and, carbon disulfide
Formula Weight 127.57
Percent Purity 98%
Chemical Name or Material 4-Chloroaniline, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause cancer.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-401-0
RTECSNumber BX0700000
TSCA TSCA
Recommended Storage The product turns dark during prolonged storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.