Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzamide, 98+%

Catalog No. AAA1380918
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1380918 50 g
AAA1380909 10 g
AAA1380930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1380918 Supplier Thermo Scientific Chemicals Supplier No. A1380918
Only null left
Add to Cart

CAS: 619-56-7 | C7H6ClNO | 155.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-56-7
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.58
MDL Number MFCD00007993
InChI Key BLNVISNJTIRAHF-UHFFFAOYSA-N
Synonym p-chlorobenzamide, benzamide, 4-chloro, 4-chloro-benzamide, p-chlorobenzoic acid amide, benzamide, p-chloro, unii-260cv6k0mr, benzamide, p-chloro-6ci,7ci,8ci, 4-chloranylbenzamide, pubchem3619, 4-chlorobenzenecarboxamide
PubChem CID 12084
IUPAC Name 4-chlorobenzamide
SMILES NC(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 176°C to 180°C
Quantity 50 g
Beilstein 774622
Formula Weight 155.58
Percent Purity ≥98%
Chemical Name or Material 4-Chlorobenzamide
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-601-1
RTECSNumber CV2443811
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.