Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzoyl-L-tryptophan Calcium Salt 98.0+%, TCI America™
SDP

Catalog No. C084925G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 56116-62-2
Molecular Formula C36H28Ca2Cl2N4O6+2
Molecular Weight (g/mol) 763.698
MDL Number MFCD00036700
InChI Key RTNAGHDBACTRBP-OLYXIJPGSA-L
PubChem CID 133126624
IUPAC Name dicalcium;(2R)-2-[(4-chlorobenzoyl)amino]-3-(1H-indol-3-yl)propanoate
SMILES C1=CC=C2C(=C1)C(=CN2)CC(C(=O)[O-])NC(=O)C3=CC=C(C=C3)Cl.C1=CC=C2C(=C1)C(=CN2)CC(C(=O)[O-])NC(=O)C3=CC=C(C=C3)Cl.[Ca+2].[Ca+2]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 723.62
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorobenzoyl-L-tryptophan Calcium Salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.