Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobutyraldehyde Dimethyl Acetal 97.0+%, TCI America™
SDP

Catalog No. C217925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C217925G 25 g
C21795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C217925G Supplier TCI America Supplier No. C217925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29882-07-3
Molecular Formula C6H13ClO2
Molecular Weight (g/mol) 152.618
MDL Number MFCD01632883
InChI Key LTLKJYMNUSSFAH-UHFFFAOYSA-N
Synonym 4-Chloro-1,1-dimethoxybutane
PubChem CID 122536
IUPAC Name 4-chloro-1,1-dimethoxybutane
SMILES COC(CCCCl)OC

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 g
UN Number 1993
Formula Weight 152.62
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorobutyraldehyde Dimethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.