Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobutyryl chloride, 98%

Catalog No. AAA122490B
Change view
Click to view available options
Quantity:
10 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA122490B 1000 g
AAA1224909 10 g
AAA1224930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA122490B Supplier Thermo Scientific Chemicals Supplier No. A122490B
Only null left
Add to Cart

CAS: 4635-59-0 | C4H6Cl2O | 140.991 g/mol

4-Chlorobutyryl chloride (4-CBCl) is mainly used in the production of pharmaceuticals and agrochemicals. It is used as pyrazinamide intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobutyryl chloride (4-CBCl) is mainly used in the production of pharmaceuticals and agrochemicals. It is used as pyrazinamide intermediate.

Solubility
soluble in diethyl ether, ethanol, slightly soluble in water.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agent, bases,
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4635-59-0
Molecular Formula C4H6Cl2O
Molecular Weight (g/mol) 140.991
MDL Number MFCD00000754
InChI Key CDIIZULDSLKBKV-UHFFFAOYSA-N
Synonym 4-chlorobutyryl chloride, butanoyl chloride, 4-chloro, 4-chlorobutanoic acid chloride, 4-chlorobutyric acid chloride, 4-chlorobutyroyl chloride, gamma-chlorobutyryl chloride, 4-chlorobutyrylchloride, gamma-chlorobutyroyl chloride, unii-3663u0vkri, butyryl chloride, 4-chloro
PubChem CID 78370
IUPAC Name 4-chlorobutanoyl chloride
SMILES C(CC(=O)Cl)CCl

Specifications

Melting Point -49°C
Density 1.258
Boiling Point 173°C to 174°C
Flash Point 85°C (185°F)
Odor Acrid
Refractive Index 1.461
Linear Formula ClCH2CH2CH2COCl
Quantity 1000 g
UN Number UN2927
Beilstein 773860
Sensitivity Moisture sensitive
Solubility Information soluble in diethyl ether,ethanol,slightly soluble in water.
Formula Weight 141
Percent Purity 98%
Chemical Name or Material 4-Chlorobutyryl chloride
Show More Show Less
Hazard Category H227-H290-H302-H314-H330-H335
Hazard Statement GHS H Statement
H301-H330-H314-H318-H227
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P234-P235-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: I
EINECSNumber 225-059-1
RTECSNumber EM1406000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.