Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorodiphenylmethane 96.0+%, TCI America™
SDP

Catalog No. C208625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C208625G 25 g
C20865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C208625G Supplier TCI America Supplier No. C208625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 831-81-2
Molecular Formula C13H11Cl
Molecular Weight (g/mol) 202.681
MDL Number MFCD00094070
InChI Key NPOGRKGIBGKRNI-UHFFFAOYSA-N
PubChem CID 13253
IUPAC Name 1-benzyl-4-chlorobenzene
SMILES C1=CC=C(C=C1)CC2=CC=C(C=C2)Cl

Specifications

Melting Point 7°C
Color Yellow
Boiling Point 153°C
Quantity 25 g
Formula Weight 202.68
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorodiphenylmethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.