Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenacyl Chloride 98.0+%, TCI America™
SDP

Catalog No. C130625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C130625G 25 g
C13065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C130625G Supplier TCI America Supplier No. C130625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 937-20-2
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.035
MDL Number MFCD00018926
InChI Key FWDFNLVLIXAOMX-UHFFFAOYSA-N
Synonym 2,4'-dichloroacetophenone, 2-chloro-1-4-chlorophenyl ethanone, 4-chlorophenacyl chloride, 2-chloro-1-4-chlorophenyl ethan-1-one, ethanone, 2-chloro-1-4-chlorophenyl, alpha,4-dichloroacetophenone, p-chlorophenacyl chloride, pubchem3372, 2,4-dichloroacetophenone, acmc-209rn9
PubChem CID 70298
IUPAC Name 2-chloro-1-(4-chlorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CCl)Cl

Specifications

Melting Point 101°C
Color White
Boiling Point 270°C
Quantity 25 g
Formula Weight 189.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorophenacyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.