Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-chlorophenol, 99+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
5kg
Packaging:
Glass bottle
Plastic drum
106-48-9
C6H5ClO
128.56
MFCD00002318
WXNZTHHGJRFXKQ-UHFFFAOYSA-N
p-chlorophenol, parachlorophenol, phenol, 4-chloro, 4-hydroxychlorobenzene, phenol, p-chloro, applied 3-78, 4-monochlorophenol, p-chlorophenic acid, p-chlorfenol, 4-chloro-1-hydroxybenzene
4684
CHEBI:28078
4-chlorophenol
C1=CC(=CC=C1O)Cl
106-48-9
C6H5ClO
128.56
MFCD00002318
WXNZTHHGJRFXKQ-UHFFFAOYSA-N
p-chlorophenol, parachlorophenol, phenol, 4-chloro, 4-hydroxychlorobenzene, phenol, p-chloro, applied 3-78, 4-monochlorophenol, p-chlorophenic acid, p-chlorfenol, 4-chloro-1-hydroxybenzene
4684
CHEBI:28078
4-chlorophenol
C1=CC(=CC=C1O)Cl
Density 1.2600g/mL
Water 0.3% max.
Quantity 100g
Specific Gravity 1.26
Melting Point 41.0°C to 45.0°C
Boiling Point 220.0°C
Viscosity 4.99 mPa.s (50°C)
Packaging Glass bottle
Flash Point 102°C
Infrared Spectrum Authentic
Color White to Yellow
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99% min. (GC)
Linear Formula ClC6H4OH
Beilstein 06,186
Merck Index 15,2157
Solubility Information Solubility in water: 2.7g/100 mL (20°C). Other solubilities: soluble in alcohol,ether,aromatic solvents,acetone,chloroform,glycerin and sodium hydroxide
Formula Weight 128.56
Physical Form Low Melting Crystalline Mass
Percent Purity 99+%
Chemical Name or Material 4-Chlorophenol
Show More Show Less

GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breath

GHS Signal Word: Warning

EINECSNumber : 203-402-6