Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenoxyacetic acid, 98%

Catalog No. p-90758
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

CAS: 122-88-3 | C8H7ClO3 | 186.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-88-3
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.59
MDL Number MFCD00004305
InChI Key SODPIMGUZLOIPE-UHFFFAOYSA-N
Synonym 4-chlorophenoxyacetic acid, 2-4-chlorophenoxy acetic acid, 4-chlorophenoxy acetic acid, 4-cpa, sure-set, tomatotone, tomato fix, acetic acid, 4-chlorophenoxy, p-chlorophenoxyacetic acid, tomato hold
PubChem CID 26229
ChEBI CHEBI:1808
IUPAC Name 2-(4-chlorophenoxy)acetic acid
SMILES C1=CC(=CC=C1OCC(=O)O)Cl

Specifications

Melting Point >156.0°C
Color White to Beige
CAS Min % 97.5
CAS Max % 100.0
Quantity 100 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Powder
Assay Percent Range 98%
Linear Formula ClC6H4OCH2CO2H
Beilstein 06, 187
Solubility Information Solubility in water: sparingly soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 186.59
Percent Purity 98%
Chemical Name or Material 4-Chlorophenoxyacetic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-581-3
RTECSNumber AG0175000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.