Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 4-Chlorophenoxyacetic acid Monoclonal Antibody (3D5)
GREENER_CHOICE

Catalog No. PIMA549742
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549742 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549742 Supplier Invitrogen™ Supplier No. MA549742
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

4-Chlorophenoxyacetic acid (4-CPA) is a synthetic auxin that is widely used as a plant growth regulator, herbicide, and hormone in agriculture. It is a white crystalline powder that is soluble in water and commonly used to control the growth of broadleaf weeds in crops like wheat, maize, and rice. 4-CPA works by mimicking the natural plant hormone indoleacetic acid (IAA) and stimulating plant growth or inhibiting it depending on the concentration used. It is also used in tissue culture to induce root formation and is being studied for its potential use in cancer therapy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 4-Chlorophenoxyacetic acid
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3D5
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of 4-D hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.