Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenoxyacetyl Chloride 98.0+%, TCI America™
SDP

Catalog No. C0832500G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
100 g
500 g

Chemical Identifiers

CAS 4122-68-3
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00000727
InChI Key VRBVHQUSAOKVDH-UHFFFAOYSA-N
Synonym 4-chlorophenoxyacetyl chloride, p-chlorophenoxyacetyl chloride, 2-4-chlorophenoxy acetyl chloride, 4-chlorophenoxy acetyl chloride, acetyl chloride, 4-chlorophenoxy, 4-chlorophenoxyacetylchloride, acmc-1aqz7, 4-chlorophenoxyacetyl chlroride, 4chlorophenoxy acetyl chloride, vrbvhqusaokvdh-uhfffaoysa
PubChem CID 77771
IUPAC Name 2-(4-chlorophenoxy)acetyl chloride
SMILES C1=CC(=CC=C1OCC(=O)Cl)Cl

Specifications

Color Yellow
Boiling Point 165°C
Quantity 500 g
UN Number 3265
Formula Weight 205.03
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 4-Chlorophenoxyacetyl Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.