Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenyl Methyl Sulfone 98.0+%, TCI America™
SDP

Catalog No. C123725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C123725G 25 g
C12375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C123725G Supplier TCI America Supplier No. C123725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 98-57-7
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.641
MDL Number MFCD00025067
InChI Key LMCOQDVJBWVNNI-UHFFFAOYSA-N
Synonym 4-chlorophenyl methyl sulfone, 1-chloro-4-methylsulfonyl benzene, p-chlorophenyl methyl sulfone, 4-methylsulfuryl chlorobenzene, benzene, 1-chloro-4-methylsulfonyl, methyl 4-chlorophenyl sulfone, sulfone, p-chlorophenyl methyl, 1-chloro-4-methylsulphonyl benzene, 4-chlorophenylmethyl sulfone, methyl p-chlorophenyl sulfone
PubChem CID 7395
IUPAC Name 1-chloro-4-methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=C(C=C1)Cl

Specifications

Melting Point 98°C
Color White
Boiling Point 60°C
Quantity 25 g
Formula Weight 190.64
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorophenyl Methyl Sulfone
EINECSNumber (3)-3469
RTECSNumber WR5730000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.