Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenylacetone 97.0+%, TCI America™
SDP

Catalog No. C252625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C252625G 25 g
C25265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C252625G Supplier TCI America Supplier No. C252625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5586-88-9
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00045214
InChI Key WEJRYKSUUFKMBC-UHFFFAOYSA-N
Synonym 4-chlorophenylacetone, 1-4-chlorophenyl acetone, 1-4-chlorophenyl propan-2-one, 1-p-chlorophenyl acetone, 1-4-chloro-phenyl-propan-2-one, p-chlorophenyl acetone, 2-propanone, 1-4-chlorophenyl, p-chlorophenylacetone, 4-chlorophenyl acetone
PubChem CID 79699
IUPAC Name 1-(4-chlorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(Cl)C=C1

Specifications

Color Yellow
Boiling Point 216°C
Quantity 25 g
Formula Weight 168.62
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorophenylacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.