Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Chlorophenylacetylene, 98%
Click to view available options
Quantity:
1 g
5 g
Description
4-Chlorophenylacetylene is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chlorophenylacetylene is used as pharmaceutical intermediate.
Solubility
Insoluble in water. Soluble in acetone, chloroform, dichloromethane, DMF, DMSO, ethanol, ethyl acetate, hexane, methanol,THF and toluene.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
4-Chlorophenylacetylene is used as pharmaceutical intermediate.
Solubility
Insoluble in water. Soluble in acetone, chloroform, dichloromethane, DMF, DMSO, ethanol, ethyl acetate, hexane, methanol,THF and toluene.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 43°C to 47°C |
| Boiling Point | 179°C |
| Quantity | 5 g |
| UN Number | UN1325 |
| Solubility Information | Insoluble in water. Soluble in acetone,chloroform,dichloromethane,DMF,DMSO,ethanol,ethyl acetate,hexane,methanol,THF and toluene. |
| Formula Weight | 136.58 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Chlorophenylacetylene |
Safety and Handling
| Hazard Category | H228-H315-H319-H335 |
| Hazard Statement | GHS H Statement H228-H319-H335 Flammable solid. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Hazard Class: 4.1; Packaging Group: II |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction