Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenylhydrazine hydrochloride, 97%

Catalog No. p-90850
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1073-70-7 | C6H7ClN2·HCl | 179.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073-70-7
Molecular Formula C6H7ClN2·HCl
Molecular Weight (g/mol) 179.05
MDL Number MFCD00012943
InChI Key YQVZREHUWCCHHX-UHFFFAOYSA-N
Synonym 4-chlorophenylhydrazine hydrochloride, 4-chlorophenyl hydrazine hydrochloride, p-chlorophenylhydrazine hydrochloride, p-chlorophenylhydrazine hcl, 4-chlorophenylhydrazine hcl, 1-4-chlorophenyl hydrazine hydrochloride, 4-chloro-phenyl-hydrazine hydrochloride, chlorophenylhydrazine 4-hcl, hydrazine, 4-chlorophenyl-, monohydrochloride, 4-chlorophenylhydrazine, chloride
PubChem CID 71600
IUPAC Name (4-chlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC=C1NN)Cl.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 203.0°C
Color Pink to White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 96.0 to 104% (Argentometry)
Packaging Glass bottle
Linear Formula NHNH2C6H4Cl·HCl
Quantity 5 g
Beilstein 15,I,106
Formula Weight 179.05
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorophenylhydrazine hydrochloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
EINECSNumber 214-030-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.