Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophthalic acid, monosodium salt, Tech., Thermo Scientific™

Catalog No. AC214195000
Click to view available options
:
500g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 56047-23-5
Infrared Spectrum Authentic
Molecular Formula C8H4ClNaO4
Linear Formula ClC6H3(CO2H)CO2Na
Quantity 500g
MDL Number MFCD00002495
Beilstein 09, II, 603
Synonym sodium 4-chlorobenzene-1,2-dicarboxylic acid
InChI Key MWZMHSGEQHVWED-UHFFFAOYSA-M
SMILES [Na+].OC(=O)C1=CC=C(Cl)C=C1C([O-])=O
IUPAC Name sodium 2-carboxy-5-chlorobenzoate
Molecular Weight (g/mol) 222.56
PubChem CID 118797044
Formula Weight 222.56
Percent Purity ≥60%
Grade Technical
Chemical Name or Material 4-Chlorophthalic acid, monosodium salt, tech.
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 259-963-2
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.