Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloropyridine-2-carboxamide, 97%

Click to view available options
Quantity:
5g
25g
100g
99586-65-9
C6H5ClN2O
156.569
MFCD01085339
XIHHOUUTBZSYJH-UHFFFAOYSA-N
4-chloropicolinamide,4-chloro-pyridine-2-carboxylic acid amide,4-chloro-2-pyridinecarboxamide,4-chlorophcolinamide,4-chloropyridine-2-carboxylic acid amide,2-pyridinecarboxamide, 4-chloro,4-chloro-pyridine-2-carboxylicacidamide,pubchem9223,acmc-209sda,ksc486q9h
303543
4-chloropyridine-2-carboxamide
C1=CN=C(C=C1Cl)C(=O)N
99586-65-9
C6H5ClN2O
156.569
MFCD01085339
XIHHOUUTBZSYJH-UHFFFAOYSA-N
4-chloropicolinamide,4-chloro-pyridine-2-carboxylic acid amide,4-chloro-2-pyridinecarboxamide,4-chlorophcolinamide,4-chloropyridine-2-carboxylic acid amide,2-pyridinecarboxamide, 4-chloro,4-chloro-pyridine-2-carboxylicacidamide,pubchem9223,acmc-209sda,ksc486q9h
303543
4-chloropyridine-2-carboxamide
C1=CN=C(C=C1Cl)C(=O)N
Color White
Quantity 5g
Melting Point 158°C to 162°C
Beilstein 115850
Formula Weight 156.57
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 4-Chloropyridine-2-carboxamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning