Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloroquinazoline, 95%, Thermo Scientific™

Catalog No. p-7227414
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Alpha Vector CHLOROQUINAZOLINE
Chemical Name or Material 4-Chloroquinazoline
CAS 5190-68-1
Melting Point 100°C to 105°C
Color Brown to White
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C8 H5 Cl N2
MDL Number 00228682
Quantity 1 g
UN Number 2811
Molecular Weight (g/mol) 164.59
Percent Purity 0.95
Physical Form Crystals or Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.