Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(+/-)-4-Chlorostyrene oxide, 97+%
Click to view available options
Quantity:
5 g
25 g
Description
(±)-4-Chlorostyrene oxide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(+/-)-4-Chlorostyrene oxide is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
(+/-)-4-Chlorostyrene oxide is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Specifications
Specifications
Density | 1.283 |
Boiling Point | 230°C |
Quantity | 5 g |
Refractive Index | 1.552 |
Solubility Information | Not miscible or difficult to mix with water. |
Formula Weight | 154.59 |
Percent Purity | 98% |
Chemical Name or Material | (+/-)-4-Chlorostyrene oxide |
Safety and Handling
missing translation for 'healthHazard1' | H315-H319 |
missing translation for 'healthHazard2' | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
missing translation for 'healthHazard3' | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
RTECSNumber | CZ0527000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction