Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 4-CMTB
GREENER_CHOICE

Catalog No. 464210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-4210 10 mg
46-425-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-4210 Supplier Tocris Bioscience™ Supplier No. 4642/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

FFA2 agonist; also exhibits positive allosteric modulatory activity

Free fatty acid receptor FFA2 agonist (pEC50 = 6.38) and positive allosteric modulator. Binds at a site distinct from the orthosteric site; modulates the activity of short-chain fatty acids at FFA2 via the FFA2 second extracellular loop (ECL2).

Chemical Identifiers

CAS 300851-67-6
Molecular Formula C14H15ClN2OS

Specifications

Quantity 10 mg
Formula Weight 294.8
Purity >98%
Molecular Weight (g/mol) 294.797
Chemical Name or Material 4-CMTB
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.