Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-pentanone 99.5%, Thermo Scientific™

Catalog No. AC127390010
Click to view available options
:
1L

Specifications

CAS 108-10-1
CAS Min % 99.4
CAS Max % 100.0
Melting Point -84.0°C
Density 0.8000g/mL
Boiling Point 117.4°C
Flash Point 14°C
Infrared Spectrum Authentic
Assay Percent Range 99.50%
Molecular Formula C6H12O
Refractive Index 1.3950 to 1.3970
Linear Formula (CH3)2CHCH2COCH3
Quantity 1 L
Beilstein 01, 691
Specific Gravity 0.8
Merck Index 15, 5253
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
SMILES CC(C)CC(=O)C
IUPAC Name 4-methylpentan-2-one
Molecular Weight (g/mol) 100.161
PubChem CID 7909
ChEBI CHEBI:82344
Vapor Pressure 21.5mbar at 20°C
Formula Weight 100.16
Percent Purity 99.5%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentanone, 99.5%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-550-1
RTECSNumber SA9275000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.